Learn More
MSC Haviland Floor Squeegee, 36 in, EPDM Blade, Tapered, 1 ea
Catalog No. 502869367
Encompass
This floor squeegee features a high-performance gray EPDM rubber blade, engineered to prevent floor marking. It offers excellent chemical resistance and maintains optimal performance across a wide temperature range, making it ideal for diverse demanding cleaning environments.
  • High-performance gray EPDM rubber blade resists marking floors
  • Excellent chemical resistance
  • Maintains properties across a wide temperature range (-60°F to 350°F)
  • Features a 14 gauge red powdered-coated steel frame
  • Engineered for superior push/pull performance
  • All steel frame with rounded corners
  • Straight frames equipped with a built-in scraper
  • Socket accepts a standard 1 1/8 in tapered handle
Catalog No. 50-286-9367 Supplier MSC Supplier No. 09318338
This floor squeegee features a high-performance gray EPDM rubber blade, engineered to prevent floor marking. It offers excellent chemical resistance and maintains optimal performance across a wide temperature range, making it ideal for diverse demanding cleaning environments.
  • High-performance gray EPDM rubber blade resists marking floors
  • Excellent chemical resistance
  • Maintains properties across a wide temperature range (-60°F to 350°F)
  • Features a 14 gauge red powdered-coated steel frame
  • Engineered for superior push/pull performance
  • All steel frame with rounded corners
  • Straight frames equipped with a built-in scraper
  • Socket accepts a standard 1 1/8 in tapered handle