Learn More
MSC Haviland Squeegee, 36 in Blade, Rubber, Tapered Handle, 1 ea
Catalog No. 502817859
Encompass
This Haviland floor squeegee features a 36 in. blade width and a rubber blade, designed with a tapered handle connection. It has a straight steel frame and comes in gray.
  • Designed for pull performance and superior push capabilities
  • Features a rubber neoprene blend blade
  • Straight frame equipped with a built-in scraper
  • Blade height is 2 in.
  • Holder material is metal
  • Double-ended design
Catalog No. 50-281-7859 Supplier MSC Supplier No. 09318635
This Haviland floor squeegee features a 36 in. blade width and a rubber blade, designed with a tapered handle connection. It has a straight steel frame and comes in gray.
  • Designed for pull performance and superior push capabilities
  • Features a rubber neoprene blend blade
  • Straight frame equipped with a built-in scraper
  • Blade height is 2 in.
  • Holder material is metal
  • Double-ended design