Learn More
Grainger Greenfield Threading Straight Flute Hand Tap, HSS, M3.5 x 0.6, Taper Chamfer, 1 ea
Catalog No. 502756616
Encompass_Preferred
This M3.5 x 0.6 straight flute hand tap is designed for precision thread cutting in a variety of metallic applications. Constructed from high-speed steel with a bright, uncoated finish, this tool provides reliable performance for creating internal threads. It features a taper chamfer style and a right-hand cutting direction, making it suitable for both through hole and blind hole applications.
  • Thread Size: M3.5 x 0.6
  • Material: High-speed steel (HSS)
  • Chamfer Type: Taper
  • Number of Flutes: 3
  • Class of Fit: 6H
  • Thread Limit: D4
  • Primary Material Application: Steel, stainless steel, aluminum, brass, bronze, copper, and cast iron
  • Drill Size: 2.9 mm
Catalog No. 50-275-6616 Supplier Grainger Supplier No. 15J388
This M3.5 x 0.6 straight flute hand tap is designed for precision thread cutting in a variety of metallic applications. Constructed from high-speed steel with a bright, uncoated finish, this tool provides reliable performance for creating internal threads. It features a taper chamfer style and a right-hand cutting direction, making it suitable for both through hole and blind hole applications.
  • Thread Size: M3.5 x 0.6
  • Material: High-speed steel (HSS)
  • Chamfer Type: Taper
  • Number of Flutes: 3
  • Class of Fit: 6H
  • Thread Limit: D4
  • Primary Material Application: Steel, stainless steel, aluminum, brass, bronze, copper, and cast iron
  • Drill Size: 2.9 mm