Learn More
Fine Science Tools Narrow Pattern Forceps, Stainless Steel, Straight, 14.5 cm
SDP
Catalog No. NC9008355
Encompass
Tip Shape: Straight/ Tips: Serrated/ Tip Dimensions: 2.25 x 1.45mm/ Alloy / Material: Stainless Steel/ Length: 14.5cm
Catalog No. NC9008355 Supplier Fine Science Tools Supplier No. 1100214
Tip Shape: Straight/ Tips: Serrated/ Tip Dimensions: 2.25 x 1.45mm/ Alloy / Material: Stainless Steel/ Length: 14.5cm