Learn More
Bellco Glass, Inc. Straight Harvesting Tube,10L 1/2"OD"
SDP
Catalog No. 501944933
Encompass_Preferred
Material: 316l stainless steal with ra finish of 32 inch or better, outside only. MADE IN THE USA. Dimensions: od x id:1/2" x .430" x 10" long
Catalog No. 50-194-4933 Supplier Bellco Glass, Inc. Supplier No. 196490085
Material: 316l stainless steal with ra finish of 32 inch or better, outside only. MADE IN THE USA. Dimensions: od x id:1/2" x .430" x 10" long