Learn More
Bellco Glass, Inc. Straight Harvesting Tube,4.0L 1/4" OD - Ra 32・Finish
SDP
Catalog No. 501944923
Encompass_Preferred
Straight Harvesting Tube for Omni Vessels. Material: 316l stainless steal with ra finish of 32 inch or better, inside and out. MADE IN THE USA. Dimensions: odxid:1/4" x 5/32" x 4.0" long
Catalog No. 50-194-4923 Supplier Bellco Glass, Inc. Supplier No. 196490025
Straight Harvesting Tube for Omni Vessels. Material: 316l stainless steal with ra finish of 32 inch or better, inside and out. MADE IN THE USA. Dimensions: odxid:1/4" x 5/32" x 4.0" long