Learn More
Glass Expansion Straight HF Resistant Adaptor MB19
SDP
Catalog No. 502703174
Encompass_Preferred
Straight HF Resistant Adaptor MB19. Compatible with Spectro (Ametek) ARCOSII SOP Blue SOP & Green SOP/DSOI Radial ICP-OES.Glass Expansion is ISO 9001:2015 certified by AB Certification, No. AU4501-9001
Catalog No. 50-270-3174 Supplier Glass Expansion Supplier No. 218093727
Straight HF Resistant Adaptor MB19. Compatible with Spectro (Ametek) ARCOSII SOP Blue SOP & Green SOP/DSOI Radial ICP-OES.Glass Expansion is ISO 9001:2015 certified by AB Certification, No. AU4501-9001