Learn More
AVANTOR MASTERFLEX (VWR LLC) STRAIGHT L/S 15/25 10/PK
Catalog No. 502742728
Encompass
Masterflex Fitting, Polypropylene, Straight, Hose Barb Union, 3/16IN ID; 10/PK, Avantor
Catalog No. 50-274-2728 Supplier AVANTOR MASTERFLEX (VWR LLC) Supplier No. MFLX3061251
Masterflex Fitting, Polypropylene, Straight, Hose Barb Union, 3/16IN ID; 10/PK, Avantor