Learn More
Grainger Straight-On Chuck Pencil Gauge 20-120psi
Catalog No. 502750811
Encompass_Preferred
The Haltec Straight-On Chuck Pencil Gauge is a precision measuring tool used to accurately measure the diameter of holes, slots, and recesses. It has a range of 0.125 to 1 inch, graduations of 0.001 inch, and an accuracy of +/- 0.001 inch. The gauge has a knurled locking ring to hold measurements and a carbide scriber tip. It can be used on materials like steel, cast iron, aluminum, plastic, and wood. This durable metal gauge is made in the USA.
Catalog No. 50-275-0811 Supplier Grainger Supplier No. 140Y04
The Haltec Straight-On Chuck Pencil Gauge is a precision measuring tool used to accurately measure the diameter of holes, slots, and recesses. It has a range of 0.125 to 1 inch, graduations of 0.001 inch, and an accuracy of +/- 0.001 inch. The gauge has a knurled locking ring to hold measurements and a carbide scriber tip. It can be used on materials like steel, cast iron, aluminum, plastic, and wood. This durable metal gauge is made in the USA.