Learn More
Glass Expansion Straight quartz connector for Agilent 7900 115 mm (for use with P/N 21-809-3982)
SDP
Catalog No. 5000556636
Encompass_Preferred
Straight quartz connector for Agilent 7900 115 mm (for use with P/N 21-809-3982) Sourced from Glass Expansion
Catalog No. 50-005-56636 Supplier Glass Expansion Supplier No. 318083615
Straight quartz connector for Agilent 7900 115 mm (for use with P/N 21-809-3982) Sourced from Glass Expansion