Learn More
Fine Science Tools Spring Scissors, 8mm Cutting Edge, Stainless Steel, 10 cm
SDP
Catalog No. NC9746586
Encompass
Tip Shape: Straight/ Tips: Sharp/ Cutting Edge: 8mm/ Tip Diameter: 0.15mm/ Alloy / Material: Stainless Steel/ Length: 10cm/ Serrated: No
Catalog No. NC9746586 Supplier Fine Science Tools Supplier No. 1502410
Tip Shape: Straight/ Tips: Sharp/ Cutting Edge: 8mm/ Tip Diameter: 0.15mm/ Alloy / Material: Stainless Steel/ Length: 10cm/ Serrated: No