Learn More
Bellco Glass, Inc. Straight Sample Pipe Assembly , 15L For 10mm Port
SDP
Catalog No. 501946491
Encompass_Preferred
For 10mm port316l sst material with electropolish finishRa finish: ra 32 or better - inside & outOverall length: 16 7/16"Insertion length: 15"Supply complete: o-ring, sst washer & sst nut. MADE IN THE USA
Catalog No. 50-194-6491 Supplier Bellco Glass, Inc. Supplier No. 790915109
For 10mm port316l sst material with electropolish finishRa finish: ra 32 or better - inside & outOverall length: 16 7/16"Insertion length: 15"Supply complete: o-ring, sst washer & sst nut. MADE IN THE USA