Learn More
Bellco Glass, Inc. Straight Sample Pipe Assembly , 3L For 10mm Port
SDP
Catalog No. 501946438
Encompass_Preferred
Straight sampling pipe for removal of small volumes of media from bioreactor vessel. Fits into 10mm port on 3L metric headplate. 316L stainless steel material with electropolish finish Ra finish 32 or better inside and out supplied with O-Ring, stainless steel washer and nut.. MADE IN THE USA
Catalog No. 50-194-6438 Supplier Bellco Glass, Inc. Supplier No. 790900027
Straight sampling pipe for removal of small volumes of media from bioreactor vessel. Fits into 10mm port on 3L metric headplate. 316L stainless steel material with electropolish finish Ra finish 32 or better inside and out supplied with O-Ring, stainless steel washer and nut.. MADE IN THE USA