Learn More
Bellco Glass, Inc. Straight Sample Pipe Assembly , 7L For 10mm Port
SDP
Catalog No. 501946458
Encompass_Preferred
Used for withdraw of contents from 7L bioreactor vessel. Fits 10mm port on 3L metric headplate. Overall Length: 13 7/16". Insertion Length: 12". Supply complete: O-Ring, SST Washer & SST nut. MADE IN THE USA
Catalog No. 50-194-6458 Supplier Bellco Glass, Inc. Supplier No. 790900072
Used for withdraw of contents from 7L bioreactor vessel. Fits 10mm port on 3L metric headplate. Overall Length: 13 7/16". Insertion Length: 12". Supply complete: O-Ring, SST Washer & SST nut. MADE IN THE USA