Learn More
Fine Science Tools Fine Scissors, CeramaCut, Ceramic Coated Stainless Steel, 9 cm
SDP
Catalog No. NC9791561
Encompass
Tip Shape: Straight/ Tips: Sharp-Sharp/ Cutting Edge: 22mm/ Alloy / Material: Ceramic Coated Stainless Steel/ Length: 9cm/ Serrated: Yes/ Feature: CeramaCut
Catalog No. NC9791561 Supplier Fine Science Tools Supplier No. 1495809
Tip Shape: Straight/ Tips: Sharp-Sharp/ Cutting Edge: 22mm/ Alloy / Material: Ceramic Coated Stainless Steel/ Length: 9cm/ Serrated: Yes/ Feature: CeramaCut