Learn More
Fine Science Tools Surgical Scissors, Tungsten Carbide, Straight, 14.5 cm
SDP
Catalog No. NC9740965
Encompass
Tip Shape: Straight/ Tips: Sharp-Sharp/ Cutting Edge: 47mm/ Alloy / Material: Stainless Steel/ Length: 14.5cm/ Serrated: No/ Feature: Tungsten Carbide
Catalog No. NC9740965 Supplier Fine Science Tools Supplier No. 1450214
Tip Shape: Straight/ Tips: Sharp-Sharp/ Cutting Edge: 47mm/ Alloy / Material: Stainless Steel/ Length: 14.5cm/ Serrated: No/ Feature: Tungsten Carbide