Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

DWK Life Sciences WHEATON™ Straight-sided Glass Jar
SDP

Catalog No. 03084160 Shop All DWK Life Sciences Products
Change view
Click to view available options
Closure Liner:
PTFE
Polyvinyl
No. per Case:
12/Case
24/Case
48/Cs.
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Closure Liner No. per Case
03-084-160 Polyvinyl 48/Cs.
03-084-159 PTFE 48/Cs.
03-084-161 PTFE 24/Case
03-084-162 PTFE 12/Case
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
Catalog No. 03-084-160 Supplier DWK Life Sciences Supplier No. W217906
Add to Cart
Edge
Add to Cart

missing translation for 'validMainframeMsg'

WHEATON™ Straight-sided Glass Jar features a wide mouth and straight smooth walls for complete content removal. Made of Type III clear borosilicate glass with white polypropylene screw caps lined in PTFE or PV. Ideal for soil sampling and environmental applications.

  • Straight smooth walls allow for complete removal of contents
  • Wide-mouth opening provides easy access and filling
  • Made of Type III clear borosilicate glass
  • White polypropylene screw caps available with PTFE or PV liner
  • Ideal for soil sampling and environmental applications
TRUSTED_SUSTAINABILITY

Specifications

Capacity (Metric) 30 mL
Color Clear
Material Soda Lime Glass
No. per Case 48/Cs.
Sterility Non-sterile
Closure Color White
Closure Included Yes
Graduated No
Mouth Wide
Closure Liner Polyvinyl
Closure Material Polypropylene
Autoclavable Not Autoclavable
Capacity (English) 1 oz.
Shape Straight-sided
Disposable Reusable
Product Type Straight-sided Jar
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.