Learn More
DWK Life Sciences Wheaton™ Straight-sided Glass Jar
SDP
Catalog No. p-7111684 Shop All DWK Life Sciences Products
Change view
Click to view available options
Closure Liner:
PTFE
Polyvinyl
No. per Case:
12/Case
24/Case
48/Case
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Closure Liner No. per Case
03-084-159 PTFE 48/Case
03-084-161 PTFE 24/Case
03-084-162 PTFE 12/Case
03-084-160 Polyvinyl 48/Case
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
Catalog No. 03-084-159 Supplier DWK Life Sciences Supplier No. W217905
Only null left
Add to Cart
Add to Cart
Straight-sided wide-mouth bottles with white polypropylene screw caps. Made of Type III clear soda lime glass.
  • Straight smooth walls allow for complete removal of contents
  • Ideal for soil sampling and environmental applications
  • With attached screw caps lined in PTFE or PV
TRUSTED_SUSTAINABILITY
Specifications
Capacity (Metric) 30 mL
Color Clear
Material Soda Lime Glass (Type 3)
No. per Case 48/Case
Closure Color White
Mouth Wide
Product Type Straight-sided Jar
Closure Liner PTFE
Closure Material Polypropylene