Learn More
Grainger Straight Union 1/2 in PK10
Catalog No. 502764334
Encompass_Preferred
The Grainger 16W606 is a Westinghouse 40-Watt T12 fluorescent light bulb that provides a cool white light at 4100K. This bulb has a medium bi-pin base and is 48 inches in length, with an average life of 20,000 hours. It has a lumen output of 2950 and a CRI of 62. The 16W606 bulb contains mercury and should be disposed of properly. Overall, this Westinghouse fluorescent tube provides bright, energy efficient lighting for commercial and industrial applications.
Catalog No. 50-276-4334 Supplier Grainger Supplier No. 16W606
The Grainger 16W606 is a Westinghouse 40-Watt T12 fluorescent light bulb that provides a cool white light at 4100K. This bulb has a medium bi-pin base and is 48 inches in length, with an average life of 20,000 hours. It has a lumen output of 2950 and a CRI of 62. The 16W606 bulb contains mercury and should be disposed of properly. Overall, this Westinghouse fluorescent tube provides bright, energy efficient lighting for commercial and industrial applications.