Learn More
Grainger Straight Union 3/8 in PK10
Catalog No. 502764795
Encompass_Preferred
The Parker 32PTC-6 is a 3/8 inch straight union made of brass with a nickel plated finish. It has a pressure rating of 3000 psi at 100°F and is designed for use with hydraulic and pneumatic systems. The union has a hex nut for tightening and O-ring seals. It connects two male threaded ports to form a removable joint. This straight union allows easy disconnection of hydraulic or pneumatic lines for maintenance while maintaining a leak-free seal when connected. It is ideal for industrial applications requiring a durable and reliable straight union.
Catalog No. 50-276-4795 Supplier Grainger Supplier No. 16W605
The Parker 32PTC-6 is a 3/8 inch straight union made of brass with a nickel plated finish. It has a pressure rating of 3000 psi at 100°F and is designed for use with hydraulic and pneumatic systems. The union has a hex nut for tightening and O-ring seals. It connects two male threaded ports to form a removable joint. This straight union allows easy disconnection of hydraulic or pneumatic lines for maintenance while maintaining a leak-free seal when connected. It is ideal for industrial applications requiring a durable and reliable straight union.