Learn More
Fine Science Tools Vannas Spring Scissors, 4mm Cutting Edge, Straight, 10.5 cm
SDP
Catalog No. NC9110622
Encompass
Tip Shape: Straight/ Tips: Sharp/ Cutting Edge: 4mm/ Tip Diameter: 0.15mm/ Alloy / Material: Stainless Steel/ Length: 10.5cm/ Serrated: No
Catalog No. NC9110622 Supplier Fine Science Tools Supplier No. 1501810
Tip Shape: Straight/ Tips: Sharp/ Cutting Edge: 4mm/ Tip Diameter: 0.15mm/ Alloy / Material: Stainless Steel/ Length: 10.5cm/ Serrated: No