Learn More
Grainger Straigt Lock-On Chuck
Catalog No. 502750885
Encompass_Preferred
The Haltec Lock-On Air Chuck securely connects to air hoses and nozzles. Made from zinc alloy, it features a 1/4 inch NPT thread, a working pressure range of 0-300 PSI, and a temperature range of -20 to 150°F. It operates with a push-to-connect and release mechanism. Suitable for impact wrenches, air drills, and blow guns, it ensures a tight seal to prevent leaks. Its compact, durable design allows easy handling and storage, making it ideal for automotive, industrial, construction, and manufacturing applications requiring secure pneumatic connections.
Catalog No. 50-275-0885 Supplier Grainger Supplier No. 140W93
The Haltec Lock-On Air Chuck securely connects to air hoses and nozzles. Made from zinc alloy, it features a 1/4 inch NPT thread, a working pressure range of 0-300 PSI, and a temperature range of -20 to 150°F. It operates with a push-to-connect and release mechanism. Suitable for impact wrenches, air drills, and blow guns, it ensures a tight seal to prevent leaks. Its compact, durable design allows easy handling and storage, making it ideal for automotive, industrial, construction, and manufacturing applications requiring secure pneumatic connections.