Learn More
MTC Bio SureStrain™ Premium Cell Strainers, 50/PK
Supplier:  MTC Bio C4100
Encompass
This item has a minimum qty of 2 per supplier requirements.
100m cell strainer, sterile, individually wrapped, w/ 1 reducing adapter
Catalog No. 50-187-1048