Learn More
Hach Company Strainer-.406 OD x 6in long
Catalog No. 5000257163 Shop All Hach Company Products
Encompass
316 Stainless steel strainer for shallow depths 6in long x 406in OD
Catalog No. 50-002-57163 Supplier Hach Company Supplier No. 2071
316 Stainless steel strainer for shallow depths 6in long x 406in OD