Learn More
Med Vet International McKesson Calculi Strainer Specimen Collector, 25-460, 100/Case
Catalog No. 503043133
Encompass_Preferred
Calculi Strainer McKesson Mesh Screen, Built-in Tab, White Plastic, Disposable All 24 Hour Specimen Collectors Mesh screen is 60 square per square inch, which is the optimal size for filtering the smallest stones while allowing unobstructed flow-through Built-in tab for easier handling and sanitary conditions Durable white plastic construction Rinsible plastic allows one patient to use strainer for several days Fits conveniently into 24-hour specimen collection containers Single patient use Disposable
Catalog No. 50-304-3133 Supplier Med Vet International Supplier No. 482530CS
Calculi Strainer McKesson Mesh Screen, Built-in Tab, White Plastic, Disposable All 24 Hour Specimen Collectors Mesh screen is 60 square per square inch, which is the optimal size for filtering the smallest stones while allowing unobstructed flow-through Built-in tab for easier handling and sanitary conditions Durable white plastic construction Rinsible plastic allows one patient to use strainer for several days Fits conveniently into 24-hour specimen collection containers Single patient use Disposable