Learn More
MTC Bio Strainer Cap for 12x75mm FlowTubes™ FACS tubes and culture tubes, with 70µm strainer mesh, opaque white, sterile, 20 bags of 25 caps, 500/pack
Catalog No. NC3345689
Encompass_Preferred
These cell strainers are ideal for detaching, isolating and straining cells from clusters and tissues. Their built-in compatibility with 12×75mm culture tubes and FACS tubes makes them especially useful for pre-sorting and preparing samples for Flourescence-activated Cell Sorting (FACS).Typically a 35µm porous mesh is used to obtain a single-cell suspension and to prevent clogging in FACS instrumentation. However, a multitude of FACS users have cited applications requiring finer pore sizes for a higher level of anti-clog protection, while others require a larger pore size for pre-sorting and/or for more efficient rapid flow.
Catalog No. NC3345689 Supplier MTC Bio Supplier No. T900970
These cell strainers are ideal for detaching, isolating and straining cells from clusters and tissues. Their built-in compatibility with 12×75mm culture tubes and FACS tubes makes them especially useful for pre-sorting and preparing samples for Flourescence-activated Cell Sorting (FACS).Typically a 35µm porous mesh is used to obtain a single-cell suspension and to prevent clogging in FACS instrumentation. However, a multitude of FACS users have cited applications requiring finer pore sizes for a higher level of anti-clog protection, while others require a larger pore size for pre-sorting and/or for more efficient rapid flow.