Learn More
TWD Scientific LLC BioX 100uM Cell Strainer. Case of 100
SDP
Catalog No. 502274279 Shop All TWD Scientific LLC Products
Encompass_Preferred
BioX 100µm sterile nylon mesh cell strainer with green polypropylene frame featuring molded tab for easy handling (compatible with biologix 50ml centrifuge tubes). Strainers come individually wrapped. 100/case, price/case
This item is not returnable. View return policy
Catalog No. 50-227-4279 Supplier TWD Scientific LLC Supplier No. BX151100
BioX 100µm sterile nylon mesh cell strainer with green polypropylene frame featuring molded tab for easy handling (compatible with biologix 50ml centrifuge tubes). Strainers come individually wrapped. 100/case, price/case