Learn More
TWD Scientific LLC BioX 40uM Cell Strainer. Case of 100
SDP
Catalog No. 502274277 Shop All TWD Scientific LLC Products
Encompass_Preferred
BioX 40µm sterile nylon mesh cell strainer with purple polypropylene frame featuring molded tab for easy handling (compatible with biologix 50ml centrifuge tubes). Strainers come individually wrapped. 100/case, price/case
This item is not returnable. View return policy
Catalog No. 50-227-4277 Supplier TWD Scientific LLC Supplier No. BX151040
BioX 40µm sterile nylon mesh cell strainer with purple polypropylene frame featuring molded tab for easy handling (compatible with biologix 50ml centrifuge tubes). Strainers come individually wrapped. 100/case, price/case