Learn More
TWD Scientific LLC BioX 70uM Cell Strainer. Case of 100
SDP
Catalog No. 502274278 Shop All TWD Scientific LLC Products
Encompass_Preferred
BioX 70µm sterile nylon mesh cell strainer with orange polypropylene frame featuring molded tab for easy handling (compatible with biologix 50ml centrifuge tubes). Strainers come individually wrapped. 100/case, price/case
This item is not returnable. View return policy
Catalog No. 50-227-4278 Supplier TWD Scientific LLC Supplier No. BX151070
BioX 70µm sterile nylon mesh cell strainer with orange polypropylene frame featuring molded tab for easy handling (compatible with biologix 50ml centrifuge tubes). Strainers come individually wrapped. 100/case, price/case