Learn More
Cayman Chemical StramustIn PhosphatSodIu 100mg
SDP
Catalog No. 502995552
Encompass_Preferred
A derivative of estradiol that destabilizes microtubules (Kds = 10 and 15 µM for MAP-1 and MAP-2, respectively); induces cell cycle arrest in mitosis at the metaphase stage in DU145 and PC3 prostate cancer cells; induces apoptosis in U87MG human malignant glioma cells
Catalog No. 50-299-5552 Supplier Cayman Chemical Supplier No. 23126100 MG
A derivative of estradiol that destabilizes microtubules (Kds = 10 and 15 µM for MAP-1 and MAP-2, respectively); induces cell cycle arrest in mitosis at the metaphase stage in DU145 and PC3 prostate cancer cells; induces apoptosis in U87MG human malignant glioma cells