Learn More
McMaster-Carr Stranded Wire, 600V AC, 22 Wire Gauge, 500 ft
Catalog No. NC1885823
Encompass
Also known as hook-up wire, this wire is primarily used for wiring electrical panels, lighting, and plant floor equipment. Stranded wire is flexible, so it’s pliable enough to be pulled around corners. The strand count is the number of strands followed by the size of the individual strand. The higher the number of strands, the more flexible the wire.
Catalog No. NC1885823 Supplier McMaster-Carr Supplier No. 8054T23
Also known as hook-up wire, this wire is primarily used for wiring electrical panels, lighting, and plant floor equipment. Stranded wire is flexible, so it’s pliable enough to be pulled around corners. The strand count is the number of strands followed by the size of the individual strand. The higher the number of strands, the more flexible the wire.