Learn More
Grainger Strap Eye Fitting Steel 3/16 in dia.
Catalog No. 502765637
Encompass_Preferred
The Grainger 16X648 is a 16-inch diameter, 6-blade, 48-inch long wooden paddle ceiling fan made by Emerson. It has an oil-rubbed bronze finish and reversible blades for summer and winter operation. This fan is UL listed and has an integrated light fixture that uses 4 candelabra base 60W bulbs (not included). It comes with a 4 speed wall control and 6-inch downrod. This is a high quality, American made ceiling fan that provides good airflow for medium to large rooms up to 400 sq ft. It has a lifetime motor warranty and 2 year component warranty.
Catalog No. 50-276-5637 Supplier Grainger Supplier No. 16X648
The Grainger 16X648 is a 16-inch diameter, 6-blade, 48-inch long wooden paddle ceiling fan made by Emerson. It has an oil-rubbed bronze finish and reversible blades for summer and winter operation. This fan is UL listed and has an integrated light fixture that uses 4 candelabra base 60W bulbs (not included). It comes with a 4 speed wall control and 6-inch downrod. This is a high quality, American made ceiling fan that provides good airflow for medium to large rooms up to 400 sq ft. It has a lifetime motor warranty and 2 year component warranty.