Learn More
Grainger Strap Eye Fitting Steel 5/32 in dia.
Catalog No. 502767978
Encompass_Preferred
The Loos Strap Eye Fitting is a 5/32 inch diameter eye fitting made of carbon steel with a zinc finish. It has a working load limit of 1,250 lbs and is rated for general purpose use. The eye opening measures 1/2 inches across. This eye fitting is designed to attach webbing straps to various hardware and equipment. It can be used for tie downs, securing tarps, creating slings, and other rigging applications. The zinc finish helps resist corrosion. This is a standard eye fitting for light to medium duty strap connection needs.
Catalog No. 50-276-7978 Supplier Grainger Supplier No. 16X647
The Loos Strap Eye Fitting is a 5/32 inch diameter eye fitting made of carbon steel with a zinc finish. It has a working load limit of 1,250 lbs and is rated for general purpose use. The eye opening measures 1/2 inches across. This eye fitting is designed to attach webbing straps to various hardware and equipment. It can be used for tie downs, securing tarps, creating slings, and other rigging applications. The zinc finish helps resist corrosion. This is a standard eye fitting for light to medium duty strap connection needs.