Learn More
Grainger Strap Fork End 304 SS Cable Size 5/32
Catalog No. 502765066
Encompass_Preferred
The LOOS Fork End Cable is a 5/32 inch cable size fork end made of galvanized steel with a zinc plating. It has a maximum working load of 1,740 lbs and is rated for 7x19 and 6x19 IWRC cable constructions. The opening width is 5/16 inches and the throat depth is 13/32 inches. This galvanized steel fork end can be used for a variety of lifting, pulling and securing applications. It meets ASME B30.26 specifications.
Catalog No. 50-276-5066 Supplier Grainger Supplier No. 16X642
The LOOS Fork End Cable is a 5/32 inch cable size fork end made of galvanized steel with a zinc plating. It has a maximum working load of 1,740 lbs and is rated for 7x19 and 6x19 IWRC cable constructions. The opening width is 5/16 inches and the throat depth is 13/32 inches. This galvanized steel fork end can be used for a variety of lifting, pulling and securing applications. It meets ASME B30.26 specifications.