Learn More
Grainger Strap Fork End 304 SS Cable Size 3/32
Catalog No. 502767071
Encompass_Preferred
The Loos Wire Rope Clevis Fitting is a 1/16" clevis fitting made of forged steel with a zinc finish. It has a working load limit of 250 lbs and is rated for general purpose use. The clevis has a 1/16" hole size and fits 1/16" wire rope. Overall length is 1-1/4" and weight is 0.8 lbs. This clevis fitting is ideal for creating an adjustable wire rope assembly and allowing the length to be changed. It can be used in a variety of light duty applications.
Catalog No. 50-276-7071 Supplier Grainger Supplier No. 16X640
The Loos Wire Rope Clevis Fitting is a 1/16" clevis fitting made of forged steel with a zinc finish. It has a working load limit of 250 lbs and is rated for general purpose use. The clevis has a 1/16" hole size and fits 1/16" wire rope. Overall length is 1-1/4" and weight is 0.8 lbs. This clevis fitting is ideal for creating an adjustable wire rope assembly and allowing the length to be changed. It can be used in a variety of light duty applications.