Learn More
Grainger Strap Fork End 304 SS Cable Size 1/8
Catalog No. 502767072
Encompass_Preferred
The Strap Fork End 304 SS Cable Size 1/8" is a stainless steel cable end fitting designed to terminate 1/8" diameter 7x7 or 7x19 stainless steel cable. It has a forked end that allows the cable to be attached to various connection points. This cable end is made from 304 stainless steel for durability and corrosion resistance. It has a working load limit of 250 lbs and is intended for light duty applications. The overall length is 1-1/4" and it will accommodate 1/8" cable. This cable end provides a finished look when terminating stainless steel cable.
Catalog No. 50-276-7072 Supplier Grainger Supplier No. 16X641
The Strap Fork End 304 SS Cable Size 1/8" is a stainless steel cable end fitting designed to terminate 1/8" diameter 7x7 or 7x19 stainless steel cable. It has a forked end that allows the cable to be attached to various connection points. This cable end is made from 304 stainless steel for durability and corrosion resistance. It has a working load limit of 250 lbs and is intended for light duty applications. The overall length is 1-1/4" and it will accommodate 1/8" cable. This cable end provides a finished look when terminating stainless steel cable.