Learn More
Grainger Strapping Buckle Regular Duty PK250
Catalog No. 502764482
Encompass_Preferred
The Strapping Buckle by Strapping is a heavy duty steel buckle that fits 5/8" polypropylene strapping. It has a working load limit of 2,650 lbs and meets military specification MIL-B-27165. The buckle is 3" long x 2-1/4" wide x 3/8" thick. It features a push down release lever to easily loosen and tighten strapping. This high quality steel buckle provides a secure hold for heavy loads.
Catalog No. 50-276-4482 Supplier Grainger Supplier No. 16P027
The Strapping Buckle by Strapping is a heavy duty steel buckle that fits 5/8" polypropylene strapping. It has a working load limit of 2,650 lbs and meets military specification MIL-B-27165. The buckle is 3" long x 2-1/4" wide x 3/8" thick. It features a push down release lever to easily loosen and tighten strapping. This high quality steel buckle provides a secure hold for heavy loads.