Learn More
Grainger Strapping Sealer Manual Standard Duty
Catalog No. 502763921
Encompass_Preferred
The Strapping Sealer from Strapack is a handheld plastic strapping tool that seals polypropylene strapping up to 1/2" wide. It features a compact ergonomic design with rubber grip handles for comfort and control. The sealer creates a strong seal using heat to melt the strapping. It has adjustable temperature control up to 400°F. The sealer requires 110V AC power and comes with a 6 foot cord. It weighs 2 lbs and measures 10" x 3" x 2.5". This is an economical and portable strapping sealer ideal for light duty packaging and bundling applications.
Catalog No. 50-276-3921 Supplier Grainger Supplier No. 16P170
The Strapping Sealer from Strapack is a handheld plastic strapping tool that seals polypropylene strapping up to 1/2" wide. It features a compact ergonomic design with rubber grip handles for comfort and control. The sealer creates a strong seal using heat to melt the strapping. It has adjustable temperature control up to 400°F. The sealer requires 110V AC power and comes with a 6 foot cord. It weighs 2 lbs and measures 10" x 3" x 2.5". This is an economical and portable strapping sealer ideal for light duty packaging and bundling applications.