Learn More
MilliporeSigma STRAT-M MEMBRANE 25MM 60/PK
Catalog No. 50727982
Encompass
Strat-M membrane is a synthetic, non-animal based model for transdermal diffusion testing that is predictive of diffusion in human skin without lot-to-lot variability, safety & storage limitations.
Catalog No. 50-727-982 Supplier MilliporeSigma Supplier No. SKBM02560
Add to Cart
Add to Cart
Strat-M membrane is a synthetic, non-animal based model for transdermal diffusion testing that is predictive of diffusion in human skin without lot-to-lot variability, safety & storage limitations.