Learn More
Agilent Technologies STRATA COOLER CRYOPRES MODULE
Catalog No. 50125551
Encompass
StrataCooler Cryo Preservation Module Convenient is ideal for controlled freezing of mammalian and insect cells. Overall survival rates is of 80-90%. Weighes 1 kg, Dimensions (WxDxH): 12 x 21 x 8 cm, Capacity: 32 Standard cryovials.
Catalog No. 50-125-551 Supplier Agilent Technologies Supplier No. 400005
StrataCooler Cryo Preservation Module Convenient is ideal for controlled freezing of mammalian and insect cells. Overall survival rates is of 80-90%. Weighes 1 kg, Dimensions (WxDxH): 12 x 21 x 8 cm, Capacity: 32 Standard cryovials.