Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Phenomenex Inc Strata-X-AW 33 µm Polymeric Weak Anion

Catalog No. 502394174 Shop All Phenomenex Products
Encompass_Preferred

Strata-X-AW 33 µm Polymeric Weak Anion 2 g / 20 mL, Giga Tubes

Catalog No. 50-239-4174 Supplier Phenomenex Inc Supplier No. 8BS038KEG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Strata-X-AW 33 µm Polymeric Weak Anion 2 g / 20 mL, Giga Tubes

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.