Learn More
Phenomenex Inc Phenomenex Strata-X-AW SPE, Polymeric Weak Anion, 33 μm, 500 mg
Catalog No. NC3553345 Shop All Phenomenex Products
Encompass_Preferred
Phenomenex Strata-X-AW 33 μm polymeric weak anion (WAX) SPE tubes are formulated for cleanup, enrichment, and preconcentration of anionic and polar analytes, including PFAS, prior to LC-MS analysis. The 33 μm particle size provides reproducible flow and high loading capacity; the 500 mg sorbent mass in a 6 mL tube is suited for environmental and routine SPE workflows.
  • 33 μm polymeric weak anion mixed-mode sorbent.
  • 500 mg sorbent mass, 6 mL tube volume.
  • Designed and tested for PFAS analysis (EPA 537.1, 533, 1633).
  • High sample loading capacity and reproducible recoveries.
  • Packaged 30 per pack for routine SPE workflows.
Catalog No. NC3553345 Supplier Phenomenex Inc Supplier No. 8BS541HCH
Phenomenex Strata-X-AW 33 μm polymeric weak anion (WAX) SPE tubes are formulated for cleanup, enrichment, and preconcentration of anionic and polar analytes, including PFAS, prior to LC-MS analysis. The 33 μm particle size provides reproducible flow and high loading capacity; the 500 mg sorbent mass in a 6 mL tube is suited for environmental and routine SPE workflows.
  • 33 μm polymeric weak anion mixed-mode sorbent.
  • 500 mg sorbent mass, 6 mL tube volume.
  • Designed and tested for PFAS analysis (EPA 537.1, 533, 1633).
  • High sample loading capacity and reproducible recoveries.
  • Packaged 30 per pack for routine SPE workflows.