Learn More
Agilent Technologies STRATACLEAN RESINSTRATACLEAN
Catalog No. 50125650
Encompass
Resin, StrataClean; Stratagene; Non-flammable slurry of proprietary hydroxylated silica; Non-toxic; For cleaning up DNA or concentrate dilute protein mixtures; Alternative to phenol chloroform extractions; 3mL
Catalog No. 50-125-650 Supplier Agilent Technologies Supplier No. 400715
Resin, StrataClean; Stratagene; Non-flammable slurry of proprietary hydroxylated silica; Non-toxic; For cleaning up DNA or concentrate dilute protein mixtures; Alternative to phenol chloroform extractions; 3mL