Learn More
Agilent Technologies STRATACLEAN RESINSTRATACLEAN
Supplier:  Agilent Technologies 400714
Encompass
StrataClean Resin 3mL (200 reactions) Nonflammable slurry of proprietary hydroxylated silica particles Rapid nontoxic alternative to/phenol/chloroform and Extracts unwanted enzymes from DNA preparations in 5 minutes
Catalog No. 50-125-649