Learn More
Agilent Technologies STRATACLONE BLUNT CLONING KIT
Catalog No. NC9627629
Encompass
New improved vector now includes amp and kan resistance. High efficiency blunt-end PCR cloning, features DNA topoisomerase I and Cre recombinase technologies with Amp and Kan resistance.
Catalog No. NC9627629 Supplier Agilent Technologies Supplier No. 240207
New improved vector now includes amp and kan resistance. High efficiency blunt-end PCR cloning, features DNA topoisomerase I and Cre recombinase technologies with Amp and Kan resistance.