Learn More
Agilent Technologies STRATACLONE PCR 20REACTIONS KT
Catalog No. NC9399561
Encompass
New improved vector now includes amp and kan resistance. High efficiency UA PCR cloning, features DNA topoisomerase I and Cre recombinase technologies.
Catalog No. NC9399561 Supplier Agilent Technologies Supplier No. 240205
New improved vector now includes amp and kan resistance. High efficiency UA PCR cloning, features DNA topoisomerase I and Cre recombinase technologies.