Learn More
Agilent Technologies STRATAGENE BENCHTOP COOLER
Supplier:  Agilent Technologies 401349
Encompass
The StrataCooler LP benchtop cooler provides the most protection for your enzymes whether in your freezer or on your benchtop. At room temperature, our cooler will maintain a temperature measuring as low as -15 deg. C even after 2 hours.
Catalog No. NC9751709