Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Phenomenex Inc 200 mg / 6 mL, Tubes

Supplier:  Phenomenex Inc 8BS100FCH

Encompass_Preferred

Strata™-X 33 m Polymeric Reversed Phase 8B-S100-FCH

Catalog No. 50-239-8425


May include imposed supplier surcharges.
Only null left
Add to Cart

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.