Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Phenomenex Inc 1 g / 12 mL, Giga Tubes Teflon

Catalog No. 502400892 Shop All Phenomenex Products
Encompass_Preferred

Strata™-X 33 m Polymeric Reversed Phase 8B-S100-JDG-T

Catalog No. 50-240-0892 Supplier Phenomenex Inc Supplier No. 8BS100JDGT
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Strata™-X 33 m Polymeric Reversed Phase 8B-S100-JDG-T

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.