Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Phenomenex Inc Strata-XL-A 100 µm Polymeric Strong Anion

Catalog No. 502397963 Shop All Phenomenex Products
Encompass_Preferred

Strata-XL-A 100 µm Polymeric Strong Anion 30 mg / 1 mL, Tubes

Catalog No. 50-239-7963 Supplier Phenomenex Inc Supplier No. 8BS053TAK
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Strata-XL-A 100 µm Polymeric Strong Anion 30 mg / 1 mL, Tubes

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.